Järn(II)klorid tetrahydrat, extra pure
Leverantör: Merck
Fara
Synonyms: Ferrous chloride tetrahydrate, Iron dichloride tetrahydrate
1.02459.9012 1.02459.1000
1.02459.9012EA 0 SEK
1.02459.9012 1.02459.1000
Järn(II)klorid tetrahydrat, extra pure
Järn(II)klorid tetrahydrat
Formel: FeCl₂·4H₂O
MW: 198,81 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00149709
CAS nummer: 13478-10-9
EINECS: 231-843-4
UN: 3260
ADR: 8,III