Capillary columns, VF-1301ms
Leverantör: VARIAN
CP9054 CP9056 CP9063 CP9064 CP9066
VARICP9054EA 21450 SEK
VARICP9054 VARICP9056 VARICP9063 VARICP9064 VARICP9066
Capillary columns, VF-1301ms
Kromatografikolonn
VF-1301ms ultra-low-bleed thin film columns have a similar selectivity to VF-624ms and are suitable for the analysis of semivolatile organic solvents, as well as PCBs and pesticides.
  • Mid polarity
  • Low bleed
  • Enhanced selectivity for USP 467 eliminates co-elution of benzene and 1,2-dichloroethane