Nimodipine, yellow powder
Leverantör: MP Biomedicals
Synonyms: 3-Isopropyl 5-(2-methoxyethyl) 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Isopropyl 2-Methoxyethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate
0215980325 0215980380
ICNA0215980325EA 820 SEK
ICNA0215980325 ICNA0215980380
Nimodipine, yellow powder
Nimodipine
Nimodipine is used in the prevention of cerebral vasospasm and resultant ischemia, a complication of subarachnoid hemorrhage.
Potent L-type Ca2+ channel antagonist.
Soluble in methanol (62,5 mg/ml); DMSO (25 mg/ml), or dioxane; insoluble in aqueous solutions and 2-hydroxypropyl-b-cyclodextrin.
Formel: C₂₁H₂₆N₂O₇
MW: 418,45 g/mol
MDL Number: MFCD00153848
CAS nummer: 66085-59-4
Order Now
Specifikation testresultat
Form Yellow Powder
Storage Store at Room Temperature (15 - 30 °C). Protect from light.