7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt, pulver
Leverantör: MP Biomedicals
Synonyms: Ammonium 7-fluoro-2,1,3-benzoxadiazole-4-sulphonate
0215808605 0215808610 0215808625
ICNA0215808605EA 2375 SEK
ICNA0215808605 ICNA0215808610 ICNA0215808625
7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt, pulver
7-Fluorobenzo-2-oxa-13-diazole-4-sulphonic acid ammonium salt
7-Fluorobenzofurazan-4-sulfonic acid ammonium salt is a sensitive and specific fluorescent labeling reagent for low molecular weight compounds and for macromolecules containing thiol groups.
It does not exhibit any fluorescence of its own and no fluorescent by-products are formed in the reaction with SBD-F. The rate of reaction of thiols with SBD-F gradually increases with increasing pH.
7-Fluorobenzofurazan-4-sulphonic acid ammonium salt is used as a fluorescent probe for thiols and pre-column labeling of biological thiols for HPLC.
Soluble in water (10 mg/1 ml) or methanol.
Formel: C₆H₆FN₃O₄S
MW: 235.189 g/mol
MDL Number: MFCD02682109
CAS nummer: 84806-27-9
Order Now
Specifikation testresultat
Identity Test Passes