Järn(III)sulfat-hydrat, powder_pale gray
Leverantör: MP Biomedicals
Varning
Synonyms: Ferric Persulphate, Diiron trissulphate hydrate, Ferric sulphate hydrate
0215804283 0215804290 0215804291
ICNA0215804283EA 535 SEK
ICNA0215804283 ICNA0215804290 ICNA0215804291
Järn(III)sulfat-hydrat, powder_pale gray
Järn(III)sulfat-hydrat
Ferric sulphate is used in dyeing as a mordant, and as a coagulant for industrial wastes. It is also used in pigments, and in pickling baths for aluminum and steel. Medically it is used as an astringent and styptic. It also acts as an important oxidative agent.
Soluble in water; sparingly soluble in alcohol; practically insoluble in acetone, ethyl acetate.
Formel: Fe₂(SO₄)₃·xH₂O
MW: 399.88 g/mol (anhy)
Smältpunkt: 1539 °C (lit.)
Storage Temperature: Omgivande
MDL Number: MFCD00149714
CAS nummer: 15244-10-7
Merck Index: 13,04065
Order Now
Specifikation testresultat
Identity Test Passes
Moisture Record Result