Benfluorex hydrochloride, powder_white
Leverantör: MP Biomedicals
0215482050 0215482010
ICNA0215482050EA 3775 SEK
ICNA0215482050 ICNA0215482010
Benfluorex hydrochloride, powder_white
Benfluorex hydrochloride
Formel: C₁₉H₂₁ClF₃NO₂
MW: 387.827 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00058493
CAS nummer: 23642-66-2
EINECS: 245-777-9