Järn(III)klorid, vattenfri, black powder
Leverantör: MP Biomedicals
Fara
Synonyms: Iron trichloride, Ferric chloride
0215349980 0215349991
ICNA0215349980EA 740 SEK
ICNA0215349980 ICNA0215349991
Järn(III)klorid, vattenfri, black powder
Järn(III)klorid
Ferric chloride is a moderately strong Lewis acid and it is used as a catalyst in organic synthesis.
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistant properties. Ferric chloride is used in sewage treatment. It is also used as a leaching agent in chloride hydrometallurgy. The ferric chloride test is a traditional colorimetric test for phenols.
Observera: Research Use Only (RUO).
Formel: FeCl₃
MW: 162.21 g/mol
Kokpunkt: ~316 °C (lit.)
Smältpunkt: 304 °C (lit.)
Densitet: 2.898 g/cm³ (25 °C) (lit.)
Storage Temperature: Omgivande
MDL Number: MFCD00011005
CAS nummer: 7705-08-0
UN: 1773
ADR: 8 III
Merck Index: 13,04048
Order Now
Specifikation testresultat
Identity Test Passes