Evans blue ~85%, black powder
Leverantör: MP Biomedicals
Fara
0215110805 0215110850
ICNA0215110805EA 500 SEK
ICNA0215110805 ICNA0215110850
Evans blue ~85%, black powder
Evans blue
Evans Blue used in double-labeling procedure for studying axonal branching.
  • Dye content: ~85%
Observera: Research Use Only (RUO).
Formel: C₃₄H₂₈N₆O₁₄S₄
MW: 960,79 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00004021
CAS nummer: 314-13-6
Order Now
Specifikation testresultat
Identity Test Passes
UV/VIS: λmax (H₂O) 601 - 611nm