Uridine-5'-diphosphate disodium salt ≥90%, white powder
Leverantör: MP Biomedicals
Synonyms: UDP sodium salt
0210120525 0210120580 0210120590 0210120591
ICNA0210120525EA 380 SEK
ICNA0210120525 ICNA0210120580 ICNA0210120590 ICNA0210120591
Uridine-5'-diphosphate disodium salt ≥90%, white powder
Uridine-5'-diphosphate disodium salt
Uridine-5'-diphosphate disodium salt is a nucleoside diphosphate.
  • P2Y receptor agonist
Uridine-5'-diphosphate disodium salt is a P2Y receptor agonist.
Soluble in water (5% w/v-clear).
Observera: Research Use Only (RUO).
Formel: C₉H₁₂N₂O₁₂P₂Na₂·×H₂O
MW: 448,124 g/mol
Storage Temperature: Frys
MDL Number: MFCD00006524
CAS nummer: 21931-53-3
Order Now
Specifikation testresultat
Identity Test Passes
Purity ≥90%