Beef extract, tan powder
Leverantör: MP Biomedicals
0210088380
0210088390
ICNA0210088380EA
2000
SEK
ICNA0210088380
ICNA0210088390
Beef extract, tan powder
Media
Basmetarial för Mikrobiologi
Beef extract is a water soluble powder, standardized for use in microbiological media.
- Soluble in water
Beef extract is used in general purpose media. It is recommended for use in bacteriological examination of water, milk, and other materials, where uniform composition of media is important and for biochemical studies, particularly fermentation reactions because of its independence from fermentable substances.