CellMaxx™ Bovine transferrin (holo) (from Serum), MP Biomedicals
Leverantör: MP Biomedicals
CellMaxx™ Bovine Transferrin is chromatographically purified from New Zealand-sourced bovine plasma in an ISO quality system assuring complete traceability and consistent high quality. Transferrin (HOLO) is an iron-rich product supplied as a heat-treated, lyophilised powder.
It is frequently used as a nutrient in cell and microbial culture and as a media supplement in bio-pharmaceutical production. Also, it can be used with a wide range of cell types.