D(-)-Luciferin sodium salt, Ultra Pure Grade
Leverantör: VWR Chemicals
Synonyms: Sodium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate, D-Luciferin, sodium, D-Luciferin sodium, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt
N456-100MGEA 0 SEK
N456-100MG N456-50MG
D(-)-Luciferin sodium salt, Ultra Pure Grade
D(-)-Luciferin sodium salt
Formel: C₁₁H₇N₂NaO₃S₂
MW: 302.30 g/mol
Storage Temperature: Frys, lågtemperatur (–40°C)
MDL Number: MFCD00044938
CAS nummer: 103404-75-7
Order Now
Specifikation testresultat
Appearance Yellow powder
Water Content (KF) ≤8%