2-(Aminomethyl)pyrazine 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Pyrazin-2-ylmethanamine
454160010 454162500
ACRO454160010EA 7500 SEK
ACRO454160010 ACRO454162500
2-(Aminomethyl)pyrazine 95%
2-(Aminomethyl)pyrazine
Formel: C₅H₇N₃
MW: 109.1292 g/mol
Kokpunkt: 87…88 °C (3 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD00673149
CAS nummer: 20010-99-5
UN: 2735
ADR: 8 III
Order Now
Specifikation testresultat
Appearance Yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥94.0 %