Bismuth(III) sulphide 99%, ren
Leverantör: Thermo Fisher Scientific
Synonyms: Dibismuth trisulphide
453991000 453995000
ACRO453991000EA 2050 SEK
ACRO453991000 ACRO453995000
Bismuth(III) sulphide 99%, ren
Bismuth(III) sulphide
Formel: Bi₂S₃
MW: 514.16 g/mol
Smältpunkt: 763
Storage Temperature: Omgivande
MDL Number: MFCD00014204
CAS nummer: 1345-07-9
Merck Index: 12,01329
Order Now
Specifikation testresultat
Appearance Grey Powder and/or chunks
Titration Complexometric 80.1 to 82.5 % (Bi)