Koppar(II)nitrat trihydrat 99%, ren
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Cupric nitrate trihydrate, Copper dinitrate trihydrate
453960010 453960050
ACRO453960010EA 1375 SEK
ACRO453960010 ACRO453960050
Koppar(II)nitrat trihydrat 99%, ren
Koppar(II)nitrat trihydrat
Formel: Cu(NO₃)₂·3H₂O
MW: 241.6 g/mol
Smältpunkt: 114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149671
CAS nummer: 10031-43-3
UN: 3085
ADR: 5.1 II
Merck Index: 13,02671
Order Now
Specifikation testresultat
Appearance Dark blue Crystalline powder or crystals
Titration Iodometric 98.5 to 104 %