Vismut(III)-oxoklorid 97%, ren
Leverantör: Thermo Fisher Scientific
Varning
453840010 453840500 453842500
ACRO453840010EA 9525 SEK
ACRO453840010 ACRO453840500 ACRO453842500
Vismut(III)-oxoklorid 97%, ren
Vismut(III)-oxoklorid
Formel: BiClO
Densitet: 7,72 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00010895
CAS nummer: 7787-59-9
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Assay 77.0 to 82.5 % (Bi)