6-Chloro-m-anisidine 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 6-Chloro-3-anisidine, 2-Chloro-5-methoxyaniline, 3-Amino-4-chloroanisole
453780050 453780250
ACRO453780050EA 1025 SEK
ACRO453780050 ACRO453780250
6-Chloro-m-anisidine 98%
6-Chloro-m-anisidine
Formel: C₇H₈ClNO
MW: 157,6 g/mol
Kokpunkt: 135…137 °C (12 mmHg)
Smältpunkt: 23…27 °C
Densitet: 1,261 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00047830
CAS nummer: 2401-24-3
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Colorless to yellow to brown Low melting solid or liquid
Infrared spectrum Conforms
GC ≥97.5 %