1-(2-Chloroethyl)-2-pyrrolidone 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-(2-chloroethyl)pyrrolidin-2-one, N-(beta-chloroethyl)-pyrrolidinone, 2-Pyrrolidinone, 1-(2-chloroethyl)-
453760010 453760050
ACRO453760010EA 1475 SEK
ACRO453760010 ACRO453760050
1-(2-Chloroethyl)-2-pyrrolidone 95%
1-(2-Chloroethyl)-2-pyrrolidone
Formel: C₆H₁₀ClNO
MW: 147,6 g/mol
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00169064
CAS nummer: 51333-90-5
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %