2-(Chloromethyl)benzoxazole 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2-(Chloromethyl)benzo[d]oxazole, 2-(Chloromethyl)-1,3-benzoxazole
453380010 453380050
ACRO453380010EA 2225 SEK
ACRO453380010 ACRO453380050
2-(Chloromethyl)benzoxazole 97%
2-(Chloromethyl)benzoxazole
Formel: C₈H₆ClNO
MW: 167.5900 g/mol
Kokpunkt: 80 °C (.1 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD05664964
CAS nummer: 41014-43-1
Order Now
Specifikation testresultat
Appearance Colorless to orange Liquid
Infrared spectrum Conforms
GC ≥94.0 %