Tetrahydropyran-2-ylmethylamine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: oxan-2-ylmethanamine, 2H-Pyran-2-methanamine, tetrahydro-, 2-Aminomethyltetrahydropyran
453370010 453370050
ACRO453370010EA 3050 SEK
ACRO453370010 ACRO453370050
Tetrahydropyran-2-ylmethylamine 97%
Tetrahydropyran-2-ylmethylamine
Formel: C₆H₁₃NO
MW: 115,18 g/mol
Kokpunkt: 110 °C (25 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD06738971
CAS nummer: 6628-83-7
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure