2-Hydroxy-6-methylbenzoic acid 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,6-Cresotic acid, 6-Hydroxy-o-toluic acid, 6-Methylsalicylic acid
453060010 453060050 453060250
ACRO453060010EA 895 SEK
ACRO453060010 ACRO453060050 ACRO453060250
2-Hydroxy-6-methylbenzoic acid 98%
2-Hydroxy-6-methylbenzoic acid
Formel: C₈H₈O₃
MW: 152.15 g/mol
Smältpunkt: 170…175 °C
Storage Temperature: Omgivande
MDL Number: MFCD01194284
CAS nummer: 567-61-3
Order Now
Specifikation testresultat
Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
Melting point 168 to 176 °C
HPLC ≥97.5 %