Dibutyl succinate 99%
Leverantör: Thermo Scientific
Synonyms: Succinic acid dibutyl ester
453010250 453011000
ACRO453010250EA 830 SEK
ACRO453010250 ACRO453011000
Dibutyl succinate 99%
Dibutyl succinate
Formel: C₁₂H₂₂O₄
MW: 230,3 g/mol
Kokpunkt: 274 °C
Smältpunkt: –29 °C
Densitet: 0,98 g/cm³
Flampunkt: 144 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040495
CAS nummer: 141-03-7
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4310 (20°C, 589 nm)
GC ≥98.5 %
Acid value ≤2.0 mg KOH/g