Trifluorothymidine 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Trifluridine, 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione, Trifluoromethyldeoxyuridine, 5-Trifluorothymidine
452910010 452912500
ACRO452910010EA 9100 SEK
ACRO452910010 ACRO452912500
Trifluorothymidine 98%
Trifluorothymidine
Formel: C₁₀H₁₁F₃N₂O₅
MW: 296,2 g/mol
Smältpunkt: 190…193 °C
Storage Temperature: Frys
MDL Number: MFCD00006534
CAS nummer: 70-00-8
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %