4-Bromo-1-methylimidazole 95%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Bromo-1-methyl-1H-imidazole
452430010 452430050
ACRO452430010EA 2150 SEK
ACRO452430010 ACRO452430050
4-Bromo-1-methylimidazole 95%
4-Bromo-1-methylimidazole
Formel: C₄H₅BrN₂
MW: 161 g/mol
Kokpunkt: 91…93 °C (,1hPa)
Flampunkt: >110 °C
Storage Temperature: Frys
MDL Number: MFCD01320501
CAS nummer: 25676-75-9
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥94.0 %