5-Bromobenzimidazole 97%

Leverantör: Thermo Fisher Scientific
Varning

Synonyms: 5-Bromo-1H-benzimidazole

451670050 451670250
ACRO451670050EA 2350 SEK
ACRO451670050 ACRO451670250
5-Bromobenzimidazole 97%
5-Bromobenzimidazole
Formel: C₇H₅BrN₂
Smältpunkt: 130…134 °C
Storage Temperature: Omgivande
MDL Number: MFCD00160001
CAS nummer: 4887-88-1

Order Now

Specifikation testresultat

Appearance Off-white to orange to brown Solid
Infrared spectrum Conforms
Melting point 129 to 132 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR