5-Aminopicolinic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid
450980010 450980050
ACRO450980010EA 1775 SEK
ACRO450980010 ACRO450980050
5-Aminopicolinic acid 98%
5-Aminopicolinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Smältpunkt: 215…217 °C
Storage Temperature: Omgivande
MDL Number: MFCD02684600
CAS nummer: 24242-20-4
Order Now
Specifikation testresultat
Appearance White to off-white to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %