6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 6-hydroxybenzo[d]thiazole-2-carbonitrile, 2-Cyano-6-hydroxybenzothiazole, 6-hydroxy-1,3-benzothiazole-2-carbonitrile
450940010 450940050
ACRO450940010EA 2300 SEK
ACRO450940010 ACRO450940050
6-Hydroxybenzo[d]thiazole-2-carbonitrile 97%
6-Hydroxybenzo[d]thiazole-2-carbonitrile
Formel: C₈H₄N₂OS
MW: 176.19 g/mol
Smältpunkt: 212…215 °C
Storage Temperature: Omgivande
MDL Number: MFCD00296905
CAS nummer: 939-69-5
Order Now
Specifikation testresultat
Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %