2,2,3,3-Tetramethoxybutane 95%
Leverantör: Thermo Fisher Scientific
Varning
450930050 450930250
ACRO450930050EA 1825 SEK
ACRO450930050 ACRO450930250
2,2,3,3-Tetramethoxybutane 95%
2,2,3,3-Tetramethoxybutane
Formel: C₈H₁₈O₄
Kokpunkt: 172…174 °C
Storage Temperature: Omgivande
MDL Number: MFCD25458126
CAS nummer: 176798-33-7
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %