Kalciumkarbonat 99%, extra pure
Leverantör: Thermo Fisher Scientific
Synonyms: Marble
450680010 450680050 450680250
ACRO450680010EA 450 SEK
ACRO450680010 ACRO450680050 ACRO450680250
Kalciumkarbonat 99%, extra pure
Kalciumkarbonat
Formel: CaCO₃
MW: 100.09 g/mol
Smältpunkt: 825 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00010906
CAS nummer: 471-34-1
Merck Index: 13,01658
Order Now
Specifikation testresultat
Appearance White to almost white Powder
Titration Complexometric 98.5 to 101.5 %
Loss on drying ≤2.0 % (1 g, 200°C)
Insoluble matter ≤0.2 % (in dilute acetic acid)