4-Phenyl-1-butyne 97%
Leverantör: Thermo Fisher Scientific
Varning
449770050 449770250
ACRO449770050EA 1925 SEK
ACRO449770050 ACRO449770250
4-Phenyl-1-butyne 97%
4-Phenyl-1-butyne
Formel: C₁₀H₁₀
Kokpunkt: 190 °C
Densitet: 0,926 g/cm³
Flampunkt: 66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041667
CAS nummer: 16520-62-0
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5210 to 1.5240 (20°C, 589 nm)
GC ≥96.0 %