Prednisolone 99%
Leverantör: Thermo Fisher Scientific
Fara
449470010 449470050 449470250
ACRO449470010EA 800 SEK
ACRO449470010 ACRO449470050 ACRO449470250
Prednisolone 99%
Prednisolone
Formel: C₂₁H₂₈O₅
Kokpunkt: 571 °C (1013 hPa)
Smältpunkt: 240 °C (dec.)
Densitet: 1,212 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003649
CAS nummer: 50-24-8
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥98.5 %
Specific optical rotation +113° to +119° (20°C, 589 nm) (c=1, ethanol)