Methyl hexanoate 99%
Leverantör: Thermo Scientific
Varning
Synonyms: Caproic acid methyl ester, Methyl caproate, Hexanoic acid methyl ester
449240010 449240500 449242500
ACRO449240010EA 2375 SEK
ACRO449240010 ACRO449240500 ACRO449242500
Methyl hexanoate 99%
Methyl hexanoate
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokpunkt: 151 °C (760 mmHg)
Smältpunkt: –71 °C
Densitet: 0,884 g/cm³
Flampunkt: 42 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009510
CAS nummer: 106-70-7
UN: 3272
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4060 (20°C, 589 nm)
GC ≥98.5 %