Dietanolamin ACS
Leverantör: Thermo Fisher Scientific
Fara
449060010 449062500
ACRO449060010EA 3200 SEK
ACRO449060010 ACRO449062500
Dietanolamin ACS
Dietanolamin
Formel: C₄H₁₁NO₂
Kokpunkt: 268 °C (760 mmHg)
Smältpunkt: 27…30 °C
Densitet: 1,0955 g/cm³
Flampunkt: 176 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002843
CAS nummer: 111-42-2
UN: 3259
ADR: 8,III
REACH: 01-2119488930-28
Order Now
Specifikation testresultat
Appearance Clear viscous liquid
Water ≤0.15 %
Sulfated ash ≤0.005 %
GC ≤1.0 % (Triethanolamine)
Color scale ≤15 APHA
Apparent equivalent weight 104.0 to 106.0