4'-Isobutylacetophenone 97%
Leverantör: Thermo Scientific
Varning
Synonyms: p-Isobutylacetophenone
448820500 448822500
ACRO448820500EA 1075 SEK
ACRO448820500 ACRO448822500
4'-Isobutylacetophenone 97%
4'-Isobutylacetophenone
Formel: C₁₂H₁₆O
MW: 176,26 g/mol
Kokpunkt: 107 °C (2 mmHg)
Densitet: 0,96 g/cm³
Flampunkt: 54 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00027393
CAS nummer: 38861-78-8
UN: 1224
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5190 (20°C, 589 nm)
GC ≥96.0 %