Natriumazid 99+% Biochemistry
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Sodium triazide
447810250 447811000 447815000
ACRO447810250EA 595 SEK
ACRO447810250 ACRO447811000 ACRO447815000
Natriumazid 99+% Biochemistry
Natriumazid
Formel: NaN₃
MW: 65.01 g/mol
Kokpunkt: 300 °C (760 mmHg)
Smältpunkt: 275 °C (dec.)
Densitet: 1.85 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00003536
CAS nummer: 26628-22-8
UN: 1687
ADR: 6.1 II
Merck Index: 13,08653
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Titration Argentometric ≥99.0 %
UV at 275 nm Cut off
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.05 % (in water)