Magnesiumklorid hexahydrat 98+%, extra_pure
Leverantör: Thermo Fisher Scientific
Synonyms: Magnesium dichloride
447700010 447700050
ACRO447700010EA 755 SEK
ACRO447700010 ACRO447700050
Magnesiumklorid hexahydrat 98+%, extra_pure
Magnesiumklorid hexahydrat
Formel: MgCl₂·6H₂O
MW: 203.3 g/mol
Kokpunkt: 1412
Smältpunkt: 117 °C
Storage Temperature: Omgivande
MDL Number: MFCD00149781
CAS nummer: 7791-18-6
Merck Index: 13,05684
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Titration Complexometric 98.0 to 101.0 %
Water 51.0 to 55.0 %
Heavy metals (as Pb) ≤10 ppm