Methyl-2,4-dichloropyrimidine-6-carboxylate 98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 2,6-Dichloro-4-pyrimidinecarboxylic acid methyl ester, 2-(3,4-dichlorophenyl)-4-methyl-5-thiazolecarboxylic acid, Methyl 2,6-dichloro-4-pyrimidinecarboxylate, Methyl 2,6-dichloropyrimidine-4-carboxylate
446580050 446580250
ACRO446580050EA 2475 SEK
ACRO446580050 ACRO446580250
Methyl-2,4-dichloropyrimidine-6-carboxylate 98%
Methyl-2,4-dichloropyrimidine-6-carboxylate
Formel: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Smältpunkt: 55…56 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01941317
CAS nummer: 6299-85-0
Order Now
Specifikation testresultat
Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
Melting point 53 to 59 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure