Kaliumnitrat 99%, ren
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Nitrate of Potash
446370025 446370100 446375000
ACRO446370025EA 1900 SEK
ACRO446370025 ACRO446370100 ACRO446375000
Kaliumnitrat 99%, ren
Potassium nitrate
Formel: KNO₃
MW: 101.1 g/mol
Kokpunkt: 400 °C (760 mmHg)
Smältpunkt: 334 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011409
CAS nummer: 7757-79-1
UN: 1486
ADR: 5.1 III
Merck Index: 13,07733
Order Now
Specifikation testresultat
Appearance White Crystalline powder and/or chunks
Loss on drying ≤0.5 %
Titration after Ion exchange ≥98.5 %
SPECIFIKATIONER
Environmentally Preferable