Kalium-tert-butanolat 25% (w/w) 2 M i 2-MeTHF (2-metyltetrahydrofuran), AcroSeal®
Leverantör: Thermo Scientific
Fara
Synonyms: Potassium tert-butylate, Potassium tert-butanolate
445831000 445838000
445831000.EA 3700 SEK
445831000. 445838000.
Kalium-tert-butanolat 25% (w/w) 2 M i 2-MeTHF (2-metyltetrahydrofuran), AcroSeal®
Kalium-tert-butanolat
Formel: C₄H₉KO
MW: 112,21 g/mol
Densitet: 0,86 g/cm³
Flampunkt: –11 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012162
CAS nummer: 865-47-4
UN: 2920
ADR: 8,I
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid (May develop some turbidity or precipitate)
Strength 1.90 to 2.20 M