1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate
445010010 445010050
ACRO445010010EA 2025 SEK
ACRO445010010 ACRO445010050
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₃H₂₁BN₂O₄
MW: 280.1278 g/mol
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD08706026
CAS nummer: 864754-16-5
Order Now
Specifikation testresultat
Appearance Colorless to yellow to orange Viscous liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure