Potassium cyclopropyltrifluoroborate 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Cyclopropyltrifluoroboric acid potassium salt
444870010 444870050
ACRO444870010EA 1725 SEK
ACRO444870010 ACRO444870050
Potassium cyclopropyltrifluoroborate 97%
Potassium cyclopropyltrifluoroborate
Formel: C₃H₅BF₃K
MW: 147.98 g/mol
Smältpunkt: 348…350 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09265031
CAS nummer: 1065010-87-8
Order Now
Specifikation testresultat
Appearance White to off-white Solid
Infrared spectrum Conforms
NMR (1H-NMR) Consistent with structure