4-Amino-3-fluorophenylboronic acid pinacol ester 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Amino-3-fluorobenzeneboronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Amino-3-fluorophenylboronic acid, pinacol ester
444500010 444500050
ACRO444500010EA 4150 SEK
ACRO444500010 ACRO444500050
4-Amino-3-fluorophenylboronic acid pinacol ester 97%
4-Amino-3-fluorophenylboronic acid pinacol ester
Formel: C₁₂H₁₇BFNO₂
MW: 237,08 g/mol
Smältpunkt: 101…102 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09033884
CAS nummer: 819058-34-9
Order Now
Specifikation testresultat
Appearance Off-white to tan Solid
Infrared spectrum Conforms
Melting point 100 to 105 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure