4-(4-Chlorophenyl)aniline 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 4'-Chloro-[1,1'-biphenyl]-4-amine, 4-Amino-4'-chlorobiphenyl
444460010 444460050
ACRO444460010EA 2375 SEK
ACRO444460010 ACRO444460050
4-(4-Chlorophenyl)aniline 97%
4-(4-Chlorophenyl)aniline
Formel: C₁₂H₁₀ClN
MW: 203,6700 g/mol
Smältpunkt: 127…131 °C
Storage Temperature: Omgivande
MDL Number: MFCD00477989
CAS nummer: 135-68-2
Order Now
Specifikation testresultat
Appearance Off-white to orange to brown Solid
Infrared spectrum Conforms
Melting point 126 to 132 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure