4-Acetylphenylboronic acid pinacol ester 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)ethanone, 4-Acetylphenylboronic acid, pinacol ester, 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
444430010 444430050
ACRO444430010EA 1250 SEK
ACRO444430010 ACRO444430050
4-Acetylphenylboronic acid pinacol ester 97%
4-Acetylphenylboronic acid pinacol ester
Formel: C₁₄H₁₉BO₃
MW: 246.1099 g/mol
Kokpunkt: 80 °C (.1Torr)
Smältpunkt: 59…62 °C
Storage Temperature: Omgivande
MDL Number: MFCD05863922
CAS nummer: 171364-81-1
Order Now
Specifikation testresultat
Appearance White to off-white Solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure