Cyclopropylacetic acid 97%
Leverantör: Thermo Scientific
Fara
443780010 443780050
ACRO443780010EA 1325 SEK
ACRO443780010 ACRO443780050
Cyclopropylacetic acid 97%
Cyclopropylacetic acid
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 60…61 °C (29 mmHg)
Densitet: 1,14 g/cm³
Flampunkt: 93 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041544
CAS nummer: 5239-82-7
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %