1-Iodo-4-(trifluoromethoxy)benzene 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-(Trifluoromethoxy)iodobenzene
443410010 443410050
ACRO443410010EA 450 SEK
ACRO443410010 ACRO443410050
1-Iodo-4-(trifluoromethoxy)benzene 97%
1-Iodo-4-(trifluoromethoxy)benzene
Formel: C₇H₄F₃IO
MW: 288.01 g/mol
Kokpunkt: 72…74 °C (15 mmHg)
Densitet: 1.84 g/cm³
Flampunkt: 78 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00042411
CAS nummer: 103962-05-6
Order Now
Specifikation testresultat
Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %