(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
Leverantör: Thermo Scientific
Varning
Synonyms: (S)-3-Fluoropyrrolidine hydrochloride
442740010 442740050
ACRO442740010EA 2325 SEK
ACRO442740010 ACRO442740050
(S)-(+)-3-Fluoropyrrolidine hydrochloride 97%
(S)-(+)-3-Fluoropyrrolidine hydrochloride
Formel: C₄H₉ClFN
MW: 125,57 g/mol
Smältpunkt: 180…182 °C
Storage Temperature: Omgivande
MDL Number: MFCD04038718
CAS nummer: 136725-53-6
Order Now
Specifikation testresultat
Appearance White to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
Specific optical rotation +5° to +11° (20°C, 589 nm) (c=1 in MeOH)