3-Bromo-5-(trifluoromethyl)aniline 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Amino-5-bromo-α,α,α-trifluorotoluene, 5-Bromo-α,α,α-trifluoro-m-toluidine, 3-Amino-5-bromobenzotrifluoride
442660050 442660250
ACRO442660050EA 920 SEK
ACRO442660050 ACRO442660250
3-Bromo-5-(trifluoromethyl)aniline 97%
3-Bromo-5-(trifluoromethyl)aniline
Formel: C₇H₅BrF₃N
Kokpunkt: 220…223 °C (760 mmHg)
Densitet: 1,697 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00236205
CAS nummer: 54962-75-3
ADR: 6.1,III
Order Now
Specifikation testresultat
Appearance Colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %