2-Amino-6-bromobenzothiazole 95%
Leverantör: Thermo Fisher Scientific
Varning
442290050 442290250
ACRO442290050EA 2050 SEK
ACRO442290050 ACRO442290250
2-Amino-6-bromobenzothiazole 95%
2-Amino-6-bromobenzothiazole
Formel: C₇H₅BrN₂S
MW: 229.1 g/mol
Smältpunkt: 212…214 °C
Storage Temperature: Omgivande
MDL Number: MFCD00152229
CAS nummer: 15864-32-1
Order Now
Specifikation testresultat
Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94.0 %