3-Thiophenemethanol 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Thienylmethanol, Thiophen-3-ylmethanol, Thiophen-3-methanol
442240050 442240250
ACRO442240050EA 1375 SEK
ACRO442240050 ACRO442240250
3-Thiophenemethanol 97%
3-Thiophenemethanol
Formel: C₅H₆OS
MW: 114.17 g/mol
Kokpunkt: 73 °C (6Torr)
Densitet: 1.211 g/cm³
Flampunkt: 100.5 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00014534
CAS nummer: 71637-34-8
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %